Learn More
Cayman Chemical S-Dibutyl 3-Hydroxybutyl Ph
SDP
Supplier:  Cayman Chemical 1478710 MG
Encompass_Preferred
An optically active isomer of TBP-OH, a compound produced from the metabolism of the organophosphorus solvent, TBP
Catalog No. 50-296-9985