Learn More
Medchemexpress LLC S-diclofenac | 912758-00-0 | 99.2% | 504.47 g/mol | C23H15Cl2NO2S3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1503510MG
Encompass_Preferred
S-diclofenac is a hybrid molecule that combines an H2S donor with the nonsteroidal anti-inflammatory scaffold of diclofenac. It shows multifaceted biological activity including activation of the p53 signaling pathway, inhibition of JNK, antioxidant effects, and anti-inflammatory activity, and has been used in both in vitro and in vivo research models to study inflammation, oxidative stress, and cardiac injury.
  • Activates the p53 signaling pathway and upregulates downstream effectors.
  • Inhibits JNK activation, modulating stress-related signaling.
  • Exhibits antioxidant and anti-inflammatory properties.
  • Demonstrates efficacy in animal models of edema and myocardial injury.
  • Provided at high listed purity to support research reproducibility.
  • Available as small solid packages and as a solution in DMSO for convenience.
  • Solid appearance described as reddish brown to red.
Catalog No. 50-002-86708