Learn More
Medchemexpress LLC S-Diclofenac | 912758-00-0 | C23H15Cl2NO2S3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY15035100MG
Encompass_Preferred
S-Diclofenac is an analytical standard and a hybrid molecule combining an H2S donor with the NSAID diclofenac. Intended for research and analytical applications, it activates the p53 signaling pathway and inhibits JNK activation. This compound demonstrates antioxidant and anti-inflammatory activities, making it suitable for studies in inflammation, cardiovascular disease, and apoptosis.
  • Used as an analytical standard for research and analytical applications
  • Activates the p53 signaling pathway
  • Inhibits JNK activation
  • Exhibits antioxidant activities
  • Exhibits anti-inflammatory activities
  • Induces apoptosis in rat aortic vascular smooth muscle cells
  • Applicable in HPLC, GC, and MS research experiments
Catalog No. 50-003-75924