Learn More
Medchemexpress LLC S-Diclofenac | 912758-00-0 | C23H15Cl2NO2S3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1503550MG
Encompass_Preferred
S-Diclofenac (ACS 15) is a hybrid molecule combining an H2S donor and the NSAID diclofenac. It activates the p53 signaling pathway, inhibits JNK activation, and exhibits antioxidant and anti-inflammatory activities.
  • Activates p53 signaling pathway
  • Inhibits JNK activation
  • Acts as an antioxidant
  • Acts as an anti-inflammatory
  • Induces p53 protein expression and apoptosis in rat aortic vascular smooth muscle cells
  • Ameliorates hindpaw edema in rat models
  • Reduces myocardial injury and cardiac dysfunction in mouse models
  • Has a purity of 99.20%
  • Appearance: reddish brown to red solid
Catalog No. 50-003-75568