Learn More
Medchemexpress LLC (S)-Donepezil | 142057-80-5 | 99.8% | C24H29NO3 | 25 MG

Supplier: Medchemexpress LLC 25MGHY14566A
(S)-Donepezil is the S-enantiomer of Donepezil, a specific and potent Acetylcholinesterase (AChE) inhibitor. It appears as a white to off-white solid and is intended for research use only.
- Specific and potent Acetylcholinesterase (AChE) inhibitor.
- Chemical formula: C24H29NO3.
- Molecular weight: 379.49.
- Soluble in DMSO (100 mg/mL).
- Recommended storage for powder: -20°C for 3 years or 4°C for 2 years.
- Recommended storage in solvent: -80°C for 6 months or -20°C for 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.