Learn More
Cayman Chemical S-EIcosapentaenyl-2-Hy 10mg
SDP
Supplier:  Cayman Chemical 900122810 MG
Encompass_Preferred
A homolog of EPEA, characterized by the addition of an (S)-β-methyl group at the terminal ethanolamine carbon
Catalog No. 50-289-2947