Learn More
Medchemexpress LLC (S)-GNE-140 | 2003234-64-6 | 99.3% | 499.04 g/mol | C25H23ClN2O3S2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY100742B100MG
Encompass_Preferred
(S)-GNE-140 is the less-active enantiomer of a lactate dehydrogenase A (LDHA) inhibitor supplied for research use. It is used as a biochemical probe to study LDHA-related metabolism and cellular lactate production. Provided as a solid with high reported purity; handle according to laboratory safety guidance.
  • Is the less-active enantiomer and inhibits lactate dehydrogenase A (LDHA).
  • Used to probe LDHA-related metabolism in biochemical and cellular studies.
  • Reported purity 99.3%.
  • Off-white to gray solid appearance.
  • Powder is stable at -20°C for long-term storage; in solvent store at -80°C for extended periods.
Catalog No. 50-002-11368