Learn More
Mitegen S-Hook Cane Storage Adapter For SKU M-CP-111-065
SDP
Supplier:  Mitegen MUP1SH1
Encompass_Preferred
S-Hook Cane Storage Adapter allows you to easily store shelved puck shipping canes (SKU M-CP-111-065) in these IC Biomedical storage dewars - TW34, TW35-6, HC34, and VHC35.
Catalog No. NC1604330