Learn More
Medchemexpress LLC HY-78131A 500mg Medchemexpress, (S)-(+)-Ibuprofen CAS:51146-56-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY78131A500MG
Encompass_Preferred
Medchemexpress, HY-78131A 500mg (S)-(+)-Ibuprofen CAS:51146-56-6 (S)-(+)-Ibuprofen is the S(+)-enantiomer of Ibuprofen that inhibits COX-1 and COX-2 activity with IC50s of 2.1 μM and 1.6 μM. (S)-(+)-Ibuprofen has analgesic, antiinflammatory and antipyretic effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0084