Learn More
Medchemexpress LLC (S)-(+)-Ibuprofen (standard) | 51146-56-6 | 99.87% | C13H18O2 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY78131AR
Encompass_Preferred
(S)-(+)-Ibuprofen (Standard) is the analytical standard of (S)-(+)-Ibuprofen, intended for research and analytical applications. It is the S(+)-enantiomer of Ibuprofen and acts as a potent COX-1 and COX-2 inhibitor with IC50s of 2.1 μM and 1.6 μM, respectively. The compound exhibits analgesic, anti-inflammatory, anticancer, and antipyretic effects. It is used as a reference standard in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard of (S)-(+)-Ibuprofen.
  • Intended for research and analytical applications.
  • S(+)-enantiomer of Ibuprofen.
  • Potent COX-1 and COX-2 inhibitor with IC50s of 2.1 μM and 1.6 μM, respectively.
  • Possesses analgesic, anti-inflammatory, anticancer and antipyretic effects.
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
Catalog No. 50-003-58142