Learn More
Medchemexpress LLC S-Indoximod (1-Methyl-L-tryptophan) | 21339-55-9 | 218.25 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN0707100MG
Encompass_Preferred
(S)-Indoximod (1-Methyl-L-tryptophan) is an inhibitor of indoleamine 2,3-dioxygenase (IDO) with a Ki of 19 μM. It can be utilized for research related to cancer and neurological disease.
  • Inhibitor of indoleamine 2,3-dioxygenase (IDO).
  • Can be used for the research of cancer.
  • Can be used for the research of neurological disease.
  • Reduces IDO activity in HEK293 cells.
  • Promotes the apoptosis of hepatic stellate cells.
  • Reverses Bacillus Calmette-Guerin (BCG)-induced neurotoxicity in mice.
  • Shows renoprotective effect in the IRI rat model.
  • Reverses cognitive, anxiety, and depressive-like behavior in bile duct ligation (BDL) rats.
Catalog No. 50-004-16203