Learn More
Medchemexpress LLC HY-B2137 100mg Medchemexpress, S-(+)-Ketoprofen CAS:22161-81-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB2137100MG
Encompass_Preferred
Medchemexpress, HY-B2137 100mg S-(+)-Ketoprofen CAS:22161-81-5 S-(+)-Ketoprofen is a potent inhibitor of both COX-1 and COX-2 with IC 50 s of 1.9 and 27 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3078