Learn More
Medchemexpress LLC HY-B0580A 5mg Medchemexpress, (S)-Ketorolac CAS:66635-92-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0580A5MG
Encompass_Preferred
Medchemexpress, HY-B0580A 5mg (S)-Ketorolac CAS:66635-92-5 (S)-Ketorolac is a nonsteroidal anti-inflammatory agent. (S)-ketorolac exhibits potent COX1 and COX2 enzyme inhibition [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2921