Learn More
Medchemexpress LLC (S)-Ketorolac | 66635-92-5 | 99.27% | 100 MG

Supplier: Medchemexpress LLC HYB0580A100MG
(S)-Ketorolac is a nonsteroidal anti-inflammatory agent exhibiting potent COX1 and COX2 enzyme inhibition. It is intended for research use only. This compound has a molecular weight of 255.27 and a chemical formula of C15H13NO3, appearing as a white to off-white solid. Shipping is recommended at room temperature in the continental US.
- Potent COX1 and COX2 enzyme inhibitor
- For research use only
- Molecular weight: 255.27
- Chemical formula: C15H13NO3
- Appearance: white to off-white solid
- Powder storage: -20°C for 3 years or 4°C for 2 years
- Solvent storage: -80°C for 2 years or -20°C for 1 year
- Solubility in DMSO: 200 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.