Learn More
Medchemexpress LLC (S)-ketorolac | 66635-92-5 | 99.3% | 255.27 g/mol | C15H13NO3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB0580A10MG
Encompass_Preferred
(S)-Ketorolac is the S-enantiomer of ketorolac, a nonsteroidal anti-inflammatory agent that potently inhibits cyclooxygenase enzymes COX-1 and COX-2. Supplied as a small-molecule research reagent, it is used in studies of prostaglandin synthesis, inflammation signaling, and COX enzyme activity.
  • High purity suitable for research uses.
  • Potent inhibitor of COX-1 and COX-2 for inflammation studies.
  • Provided as a solid small-molecule reagent with formula C15H13NO3.
  • Molecular weight 255.27 g/mol.
  • Documentation available: SDS, COA, and analytical data.
  • Available in small pack sizes including 10 mg.
Catalog No. 50-002-98021