Learn More
Cayman Chemical S-LInoleyl-2-Hydroxy-1 10mg
SDP
Supplier:  Cayman Chemical 900123510 MG
Encompass_Preferred
A homolog of LOEA, characterized by the addition of an (S)-β-methyl group at the terminal ethanolamine carbon
Catalog No. 50-289-3655