Learn More
Medchemexpress LLC (S)-Mephenytoin (Standard) | 70989-04-7 | 99.9% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB1184AR5MG
Encompass_Preferred
(S)-Mephenytoin (Standard) is the analytical standard of (S)-Mephenytoin intended for research and analytical applications. This anticonvulsive agent is a substrate of the cytochrome P450 (CYP) isoform CYP2C19 and can be used for the analysis of cytochrome P450 metabolism.
  • Reference standard supplied for assay
  • Used in qualitative research experiments in HPLC, GC and MS
  • Used in quantitative research experiments in HPLC, GC and MS
  • Used in methodological research experiments in HPLC, GC and MS
Catalog No. 50-004-11286