Learn More
Medchemexpress LLC (S)-Mephenytoin (Standard) | 70989-04-7 | MFCD00270025 | 99.9% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB1184AR
Encompass_Preferred
(S)-Mephenytoin (Standard) is the analytical standard of (S)-Mephenytoin. This product is intended for research and analytical applications. (S)-Mephenytoin ((+)-Mephenytoin) is an anticonvulsive agent and a substrate of the cytochrome P450 (CYP) isoform CYP2C19. It can be used for the analysis of cytochrome P450 metabolism.
  • Analytical standard
  • Intended for research and analytical applications
  • Anticonvulsive agent
  • Substrate of cytochrome P450 (CYP) isoform CYP2C19
  • Can be used for the analysis of cytochrome P450 metabolism
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-13931