Learn More
Cayman Chemical S-Methopren 100mg
SDP
Supplier:  Cayman Chemical 16807100 MG
Encompass_Preferred
A widely-used insect growth regulator; acts as an activator of the juvenile hormone receptor, known as Methoprene-tolerant, or Met (Kd = 12.3 nM)
Catalog No. 50-298-0593