Learn More
TARGETMOL CHEMICALS INC S-methyl-KE-298 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T42821MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes.S-methyl-KE-298 is the second main metabolite in plasma,is a methyl conjugate of deacetyl-KE-298.KE-298 inhibits matrix metalloproteinase (MMP-1) production from rheumatoid arthritis (RA) synovial cells. Purity 96.9%
Catalog No. 50-287-8768