Learn More
Cayman Chemical S-methyl-L-ThIocItrulInhy 10mg
SDP
Supplier:  Cayman Chemical 8058510 MG
Encompass_Preferred
S-methyl-L-Thiocitrulline is a potent NOS inhibitor with selectivity toward the neuronal isoform compared to eNOS and iNOS. For human enzymes, it exhibits Ki values of 1.2, 11, and 40 nM for nNOS, eNOS, and iNOS, respectively.{1325,1293}
Catalog No. 50-289-5356