Learn More
Medchemexpress LLC S-Metolachor 10mM 1mL | 87392-12-9 | C15H22ClNO2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11727910MM 1ML
Encompass_Preferred
(S)-Metolachlor is the S-enantiomer of the chloroacetanilide herbicide metolachlor, supplied as a 10 mM solution in DMSO (1 mL) for research and analytical applications. The substance is listed under CAS 87392-12-9 and is reported with 99.09% purity.
  • Supplied as 10 mM solution in DMSO, 1 mL.
  • High purity (99.09%), suitable for analytical work.
  • Useful for pesticide, agrochemical, and environmental research.
  • COA, SDS, and product datasheet available for quality and safety information.
  • Also offered in solid pack sizes for method development and standards.
Catalog No. 50-002-28566