Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-Mirtazapine | 61337-87-9 | 99.8% | 5 MG
SDP

Supplier:  Medchemexpress LLC HYB0352A5MG

Encompass_Preferred

(S)-Mirtazapine ((S)-Org3770) is an S(+)-enantiomer of Mirtazapine, exhibiting pronociceptive properties within an animal model of acute thermal nociception. It functions as a stereoselective 5-HT2 receptor antagonist and undergoes metabolism by CYP2D6 and CYP1A2.

  • It is an S(+)-enantiomer of Mirtazapine.
  • It has pronociceptive properties in an animal model of acute thermal nociception.
  • It is a stereoselective 5-HT2 receptor antagonist.
  • It is metabolized by CYP2D6 and CYP1A2.
  • The molecular weight is 265.35.
  • The chemical formula is C17H19N3.
  • It appears as a solid.
  • The color ranges from off-white to light yellow.
  • The purity is 99.76%.
  • The enantiomeric excess is 98.35%.

Catalog No. 50-004-05317


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.