Learn More
Medchemexpress LLC (S)-N-(1H-Indole-3-acetyl)tryptophan | 57105-53-0 | 99.9% | 361.39 | 25 MG

Supplier: Medchemexpress LLC 25MGHY138815
(S)-N-(1H-Indole-3-acetyl)tryptophan is a tryptophan derivative known for its weak inhibition of β-D-glucosidase. This chemical is provided as a white to off-white solid and is intended strictly for research applications, not for patient use.
- Purity of 99.87%
- Molecular formula C21H19N3O3
- Soluble in DMSO
- Weakly inhibits β-D-glucosidase
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.