Learn More
Medchemexpress LLC S-n-Butyrylthiocholine iodide | 1866-16-6 | 99.7% | 25 G
Supplier: Medchemexpress LLC 25GHYD0905
S-n-Butyrylthiocholine iodide is a substrate of BChE and is commonly used for in vitro enzymatic experiments.
- Substrate of BChE.
- Used for in vitro enzymatic experiments.
- Targeted for biochemical assay reagents.
- Pathway is others.