Learn More
STA PHARMACEUTICAL US LLC (S)-N-Fmoc-alpha-(4-pentenyl)alanine | 1 g | CAS 288617-73-2 | MDL MFCD09879888
Supplier: STA PHARMACEUTICAL US LLC STAAA110821GM
(S)-N-Fmoc-alpha-(4-pentenyl)alanine is a Amino Acid reagent (Subcategory: Alkenyl AA) sold by WuXi TIDES. Offered in 1 g. Store at 4 °C. SDS available for reference.
Specifications
- CAS: 288617-73-2
- MDL: MFCD09879888
- InChIKey: MRJFPZWLOJOINV-QHCPKHFHSA-N
- Molecular Weight: 379.456
- Molecular Formula: C23H25NO4
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 7.8 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (S)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-2-methylhept-6-enoic acid
- SMILES: C=CCCC[C@@](NC(OCC1C2=CC=CC=C2C3=CC=CC=C31)=O)(C)C(O)=O
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.