Learn More
Cayman Chemical S-NItroso-L-glutathIon 50mg
SDP
Supplier:  Cayman Chemical 8224050 MG
Encompass_Preferred
S-Nitroso-L-glutathione is an S-nitrosothiol NO donor based on naturally-occurring glutathione. It is a potent smooth muscle relaxant and inhibitor of platelet aggregation. Its stability in solution varies from seconds to hours depending on temperature, buffer composition, and metal content.{1277,5064,4274,3632}
Catalog No. 50-289-3109