Learn More
Medchemexpress LLC S-nornicotine | 494-97-3 | 99.3% | 148.21 g/mol | C9H12N2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW04043010MM/1ML
Encompass_Preferred
(S)-Nornicotine is the S-enantiomer of nornicotine, an endogenous metabolite of nicotine supplied as a ready-to-use stock solution for research and analytical applications. The product is commonly provided as a 10 mM solution in DMSO (1 mL) and is intended for laboratory use only; safety documentation and a certificate of analysis are available.
Catalog No. 50-002-09340