Learn More
Hampton Research S.O. PHOSPHATE #02 PH 5.2
SDP
Supplier:  Hampton Research HR225102
Encompass
HR2-251-02/StockOptions Phosphate #02, 1.0 M Sodium phosphate monobasic monohydrate/Potassium phosphate dibasic pH 5.2 (0.976 M Sodium phosphate monobasic monohydrate, 0.024 M Potassium phosphate dibasic), 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-169