Learn More
Hampton Research S.O. PHOSPHATE #04 PH 5.6
SDP
Supplier:  Hampton Research HR225104
Encompass
HR2-251-04/StockOptions Phosphate #04, 1.0 M Sodium phosphate monobasic monohydrate/Potassium phosphate dibasic pH 5.6 (0.900 M Sodium phosphate monobasic monohydrate, 0.100 M Potassium phosphate dibasic), 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-171