Learn More
Hampton Research S.O. PHOSPHATE #09 PH 6.6
SDP
Supplier:  Hampton Research HR225109
Encompass
HR2-251-09/StockOptions Phosphate #09, 1.0 M Sodium phosphate monobasic monohydrate/Potassium phosphate dibasic pH 6.6 (0.504 M Sodium phosphate monobasic monohydrate, 0.496 M Potassium phosphate dibasic), 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-176