Learn More
Hampton Research S.O. PHOSPHATE #17 PH 8.2
SDP
Supplier:  Hampton Research HR225117
Encompass
HR2-251-17/StockOptions Phosphate #17, 1.0 M Sodium phosphate monobasic monohydrate/Potassium phosphate dibasic pH 8.2 (0.040 M Sodium phosphate monobasic monohydrate, 0.960 M Potassium phosphate dibasic), 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-184