Learn More
Medchemexpress LLC S-oxiracetam | 88929-35-5 | MFCD00242951 | 99.6% | 158.16 g/mol | C6H10N2O3 | 100MG
SDP
Supplier:  Medchemexpress LLC HY126049100MG
Encompass_Preferred
S-oxiracetam is the S-enantiomer of oxiracetam, a racetam-family nootropic research compound used in neuroscience and neuroprotection studies. Supplied as a high-purity research chemical in solid and solution formats for laboratory applications.
  • High purity suitable for analytical and in vitro research.
  • Available in multiple pack sizes, including solid and DMSO solution formats.
  • Recommended storage: sealed at 4°C; specific solvent storage guidelines apply.
  • Characterized by CAS 88929-35-5 and molecular weight 158.16 g/mol.
  • Synonyms include (S)-(-)-oxiracetam and (S)-ISF2522.
Catalog No. 50-002-36223