Learn More
Medchemexpress LLC S-(p-Nitrobenzyl)glutathione | 6803-19-6 | 98.99% | 442.44 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY148208100MG
Encompass_Preferred
S-(p-Nitrobenzyl)glutathione is a competitive glutathionase inhibitor. It is converted to corresponding cysteine derivatives by rat kidney microsomes and can be used for research on the metabolic breakdown of glutathione by the glutathionase system.
  • Competitive glutathionase inhibitor
  • Converted to corresponding cysteine derivatives by rat kidney microsomes
  • Used for research on metabolic breakdown of glutathione by the glutathionase system
  • Amino acid derivative
Catalog No. 50-003-63658