Learn More
Medchemexpress LLC S-(p-Nitrobenzyl)glutathione | 6803-19-6 | 99.0% | 442.44 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY14820810MM/1ML
Encompass_Preferred
S-(p-Nitrobenzyl)glutathione is a competitive glutathionase inhibitor that is converted to corresponding cysteine derivatives by rat kidney microsomes. It is used for research on the metabolic breakdown of glutathione by the glutathionase system. In solvent, store at -80°C for 6 months or -20°C for 1 month.
  • Competitive glutathionase inhibitor
  • Converted to cysteine derivatives by rat kidney microsomes
  • Used for metabolic breakdown research
  • Solution storage: -80°C for 6 months, -20°C for 1 month
Catalog No. 50-003-63729