Learn More
Medchemexpress LLC S-(p-Nitrobenzyl)glutathione | 6803-19-6 | 99.0% | 442.44 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1482085MG
Encompass_Preferred
S-(p-Nitrobenzyl)glutathione is a competitive glutathionase inhibitor that is converted to corresponding cysteine derivatives by rat kidney microsomes. It can be used for research on the metabolic breakdown of glutathione by the glutathionase system.
  • Competitive glutathionase inhibitor
  • Converted to cysteine derivatives by rat kidney microsomes
  • Used for research on the metabolic breakdown of glutathione by the glutathionase system
Catalog No. 50-003-63164