Learn More
Medchemexpress LLC S-(p-Nitrobenzyl)glutathione | 6803-19-6 | 99.0% | 442.44 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14820850MG
Encompass_Preferred
S-(p-Nitrobenzyl)glutathione is a competitive glutathionase inhibitor. It is converted to the corresponding cysteine derivatives by rat kidney microsomes and can be used for research into the metabolic breakdown of glutathione by the glutathionase system.
  • Competitive glutathionase inhibitor
  • Converted to cysteine derivatives by rat kidney microsomes
  • Useful for research into glutathione metabolic breakdown
Catalog No. 50-003-63079