Learn More
Medchemexpress LLC S-(p-Nitrobenzyl)glutathione | 6803-19-6 | 99.0% | 442.44 g/mol | C17H22N4O8S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY14820810MG
Encompass_Preferred
S-(p-Nitrobenzyl)glutathione is a glutathione derivative used as a competitive glutathionase inhibitor in biochemical and enzymatic research. The compound is supplied as a white to off-white solid, with high purity and characterized by CAS 6803-19-6, molecular formula C17H22N4O8S, and molecular weight 442.44 g/mol.
  • Competitive glutathionase inhibitor suitable for enzymology studies.
  • High purity (~99%) appropriate for biochemical assays.
  • White to off-white solid, easy to weigh and dissolve for experiments.
  • Available as powder and as a ready-to-use 10 mM solution in DMSO for screening.
  • Stable when stored as powder at -20°C; solvent solutions require low-temperature storage.
  • Accompanied by analytical and safety documents (datasheet, COA, SDS) for traceability.
Catalog No. 50-002-86556