Learn More
Cayman Chemical S-p38 MAPK InhIbItr III 500ug
SDP
Supplier:  Cayman Chemical 19696500 UG
Encompass_Preferred
A methylsulfanylimidazole that inhibits p38 MAP kinase (IC50 = 0.90 µM in vitro); potently blocks the release of TNF-α and IL-1β from human peripheral blood mononuclear cells (IC50s = 0.37 and 0.044 µM, respectively)
Catalog No. 50-298-4507