Learn More
Medchemexpress LLC (S)-(-)-Perillic acid | 23635-14-5 | 95.0% | 166.22 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY113471A10MM/1ML
Encompass_Preferred
(S)-(-)-Perillic acid is a terpenoid plant extract that exhibits antimicrobial and anticancer properties. It functions as an apoptosis inducer, leading to cell cycle arrest and an increase in the levels of Bax, Bcl2, p21, and caspase-3 proteins. This product is suitable for cancer and infection research.
  • Demonstrated capabilities against microbes and cancer cells.
  • Induces cell apoptosis and cell cycle arrest.
  • Increases the levels of Bax, Bcl2, p21, and caspase-3 proteins.
  • Can be utilized for cancer and infection research.
Catalog No. 50-004-36711