Learn More
Medchemexpress LLC (S)-(-)-Perillic acid | 23635-14-5 | 95.0% | 166.22 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY113471A100MG
Encompass_Preferred
(S)-(-)-Perillic acid is a terpenoid plant extract known for its antimicrobial and anticancer properties. It promotes cell apoptosis and cell cycle arrest, making it valuable for research in cancer and infection.
  • Inhibits growth of arterial smooth muscle cells.
  • Blocks cell cycle progression in G0/G1 interphase.
  • Induces apoptosis in smooth muscle cells.
  • Increases levels of Bax, Bcl2, p21, and caspase-3 proteins.
  • Inhibits long-term proliferation of A549 cells.
Catalog No. 50-004-37530