Learn More
Medchemexpress LLC (S)-(-)-Perillyl alcohol | 18457-55-1 | 98.0% | 152.24 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1165141G
Encompass_Preferred
(S)-(-)-Perillyl alcohol is a monoterpene found in lavender that inhibits farnesylation of Ras, upregulates the mannose-6-phosphate receptor, and induces apoptosis. It exhibits anti-cancer activity. For research use only; not for sale to patients.
  • Inhibits farnesylation of Ras
  • Upregulates the mannose-6-phosphate receptor
  • Induces apoptosis
  • Anti-cancer activity
Catalog No. 50-004-37815