Learn More
Cayman Chemical S-PhenylsulfonylcysteIn 10mg
SDP
Supplier:  Cayman Chemical 1928110 MG
Encompass_Preferred
A reagent for blocking thiol (-SH) groups on proteins and peptides, as a first step of the thiosulfonate switch technique
Catalog No. 50-298-3758