Learn More
Meso Scale Discovery S-Plex human-IL-10 kit - 5 plate kit
SDP
Supplier:  Meso Scale Discovery K151Y2S2
Encompass
S-PLEX is MSD’s ultra-sensitive assay platform. It can dramatically improve the sensitivity of immunoassays, reducing the lower limit of detection (LLOD) by 10- to 1000-fold over other assay methods. The S-PLEX Human IL-10 Assay has the essential components for an ultrasensitive assay that measures IL-10 in a single-analyte format.
Catalog No. NC2868733