Learn More
Meso Scale Discovery S-PLEX Human CTLA-4 KitQP
SDP
Supplier:  Meso Scale Discovery K151AGLS21
Encompass
The Meso Scale Discovery S-PLEX Human CTLA-4 Kit is a multiplex immunoassay for measuring CTLA-4 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity a wide dynamic range and low sample use The assay needs 50 L of sample and is compatible with MSD instruments The kit includes a 96-well plate with capture antibodies detection antibodies Read Buffer Diluent 2 and calibrators and controls for standard curve generation and assay validation It has high specificity and 1% cross-reactivity enabling rapid accurate CTLA-4 quantification for research
Catalog No. 50-000-80330