Learn More
Meso Scale Discovery S-PLEX Human G-CSF Kit
SDP
Supplier:  Meso Scale Discovery K151K3S2
Encompass
The S-PLEX Human G-CSF Kit from Meso Scale Diagnostics is a multiplex immunoassay kit for the quantitative measurement of granulocyte colony-stimulating factor (G-CSF) in human serum plasma and cell culture supernatants The kit utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer calibrator and control The assay can detect G-CSF concentrations from 2 4-10 000 pg/mL and has high specificity with minimal cross-reactivity
Catalog No. 50-000-82340