Learn More
Meso Scale Discovery S-PLEX Human GM-CSF Kit, Kit Size 5 each, Sensitivity 1.9 fg/mL, Dynamic Range 1.9 - 26,000 fg/mL, Plate Type 96-well, Electrochemiluminescence technology

Supplier: Meso Scale Discovery K151F3S2
The MESO SCALE DISCOVERY S-PLEX Human GM-CSF Kit is designed for the sensitive detection and quantification of Human Granulocyte-Macrophage Colony Stimulating Factor (GM-CSF) in biological samples. Utilizing advanced ECL technology, this kit enhances sensitivity dramatically, allowing for the measurement of low-abundance proteins. It's ideal for research in cytokines, chemokines, and immunology/inflammation.
- Assay platform utilizing ultra-sensitive ECL technology
- Detects GM-CSF with a sensitivity of 1.9 fg/mL
- Dynamic range spanning from 1.9 to 26,000 fg/mL
- Compatible with 96-well plates
- Supports various sample types, including serum and plasma
- Mouse monoclonal antibodies for capture and detection
- Recommended storage conditions for multi-component kits
- Gene ID: 1437, UniProt ID: P04141
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.