Learn More
Meso Scale Discovery S-PLEX Human IFN- 2a KitQP
SDP
Supplier:  Meso Scale Discovery K151P3S21
Encompass
The S-PLEX Human IFN alpha2a Kit from Meso Scale Discovery is an immunoassay kit for the quantitative measurement of human interferon alpha2a (IFN 2a) in serum plasma cell culture supernatants cell lysates and tissue homogenates The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and rapid results The assay requires 50 L or less sample volume and can be read on MSD instrument platforms The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and positive control
Catalog No. 50-000-82330