Learn More
Meso Scale Discovery S-PLEX Human IFN-B KitQP
SDP
Supplier:  Meso Scale Discovery K151ADRS21
Encompass
The S-PLEX Human IFN beta Kit by Meso Scale Discovery is a highly sensitive immunoassay that quantitatively measures human interferon beta (IFN- ) in serum plasma cell culture supernatant and other biological matrices The assay utilizes electrochemiluminescence (ECL) detection technology and provides accurate reproducible results in just a few hours This kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer sample diluent calibrator and control The lower limit of detection is 0 08 pg/mL The S-PLEX Human IFN beta Kit enables researchers to reliably quantify IFN- levels for the study of multiple sclerosis and other diseases
Catalog No. 50-000-79574