Learn More
Meso Scale Discovery S-PLEX Human IL-10 Kit 25 PL
SDP
Supplier:  Meso Scale Discovery K151Y2S4
Encompass
The Meso Scale Discovery S-PLEX Human IL-10 Kit is a multiplex immunoassay that allows simultaneous measurement of IL-10 in human serum plasma and cell culture supernatants The assay uses electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG Read Buffer Diluent 2 Calibrator Blend and sample types validated include serum plasma and cell culture supernatant The assay has high sensitivity with an LLOQ of 0 07 pg/mL and upper limit of 10 000 pg/mL
Catalog No. 50-000-79712
Only null left
Add to Cart
Request a Quote